Assay Details
Gene Symbol: SYNGR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046436.2 | NP_001039901.1 | 1 - 2 | 85 | 97 | |
GenBank mRNA | BC110272.1 | - | 1 - 2 | 88 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC110272.1, NM_001046436.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
