Assay Details
| Gene Symbol: ELOC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046493.2 | NP_001039958.1 | 1 - 2 | 133 | 59 | |
| GenBank mRNA | BC112626.1 | - | 1 - 2 | 90 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC112626.1, NM_001046493.2 transcripts.
Set Membership: |
|

ELOC
Genomic Map