Assay Details
| Gene Symbol: ELOC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046493.2 | NP_001039958.1 | 2 - 3 | 187 | 71 | |
| NM_001359989.1 | NP_001346918.1 | 2 - 3 | 265 | 71 | ||
| GenBank mRNA | BC112626.1 | - | 2 - 3 | 144 | 71 | |
| EE218883.1 | - | 2 - 3 | 183 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC112626.1, NM_001046493.2, NM_001359989.1 transcripts.
Set Membership: |
|

ELOC
Genomic Map