Assay Details
| Gene Symbol: ELOC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046493.2 | NP_001039958.1 | 3 - 4 | 339 | 107 | |
| NM_001359989.1 | NP_001346918.1 | 3 - 4 | 417 | 107 | ||
| GenBank mRNA | BC112626.1 | - | 3 - 4 | 296 | 107 | |
| EE218883.1 | - | 3 - 4 | 335 | 107 | ||
More Information
Set Membership: |
|

ELOC
Genomic Map