Assay Details
| Gene Symbol: PDZK1IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046521.2 | NP_001039986.1 | 1 - 2 | 194 | 126 | |
| GenBank mRNA | BC112562.1 | - | 1 - 2 | 195 | 126 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC112562.1, NM_001046521.2 transcripts.
Set Membership: |
|

PDZK1IP1
Genomic Map