Assay Details
| Gene Symbol: VSIG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046529.2 | NP_001039994.1 | 4 - 5 | 632 | 109 | |
| GenBank mRNA | BC112495.1 | - | 4 - 5 | 633 | 109 | |
More Information
Set Membership: |
|

VSIG4
Genomic Map