Assay Details
| Gene Symbol: NARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046572.2 | NP_001040037.1 | 12 - 13 | 1365 | 104 | |
| GenBank mRNA | BC105358.1 | - | 12 - 13 | 1366 | 104 | |
More Information
Set Membership: |
|

NARS1
Genomic Map