Assay Details
Gene Symbol: MRPS24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046601.2 | NP_001040066.1 | 2 - 3 | 121 | 88 | |
GenBank mRNA | BC111632.1 | - | 2 - 3 | 122 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
