Assay Details
Gene Symbol: PFN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075377.2 | NP_001068845.1 | 1 - 2 | 269 | 72 | |
GenBank mRNA | BC111657.1 | - | 1 - 2 | 270 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC111657.1, NM_001075377.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
