Assay Details
| Gene Symbol: PFN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075377.2 | NP_001068845.1 | 3 - 4 | 533 | 69 | |
| GenBank mRNA | BC111657.1 | - | 3 - 4 | 534 | 69 | |
More Information
Set Membership: |
|

PFN4
Genomic Map