Assay Details
| Gene Symbol: C5H12orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075525.1 | NP_001068993.1 | 1 - 2 | 381 | 123 | |
| GenBank mRNA | BC110027.1 | - | 1 - 2 | 381 | 123 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC110027.1, NM_001075525.1 transcripts.
Set Membership: |
|

C5H12orf57
Genomic Map