Assay Details
Gene Symbol: MPHOSPH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075564.2 | NP_001069032.1 | 2 - 3 | 193 | 147 | |
GenBank mRNA | BC110128.1 | - | 2 - 3 | 194 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
