Assay Details
Gene Symbol: WDR53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075772.2 | NP_001069240.1 | 2 - 3 | 550 | 62 | |
GenBank mRNA | BC109979.1 | - | 2 - 3 | 551 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
