Assay Details
| Gene Symbol: ACRV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075926.2 | NP_001069394.1 | 1 - 2 | 150 | 73 | |
| GenBank mRNA | BC109966.1 | - | 1 - 2 | 152 | 73 | |
More Information
Set Membership: |
|

ACRV1
Genomic Map