Assay Details
| Gene Symbol: ACRV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075926.2 | NP_001069394.1 | 3 - 4 | 798 | 65 | |
| GenBank mRNA | BC109966.1 | - | 3 - 4 | 800 | 65 | |
More Information
Set Membership: |
|

ACRV1
Genomic Map