Assay Details
| Gene Symbol: DRAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076214.1 | NP_001069682.1 | 6 - 7 | 619 | 100 | |
| GenBank mRNA | BC110127.1 | - | 6 - 7 | 619 | 100 | |
More Information
Set Membership: |
|

DRAP1
Genomic Map