Assay Details
| Gene Symbol: APOD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076301.2 | NP_001069769.1 | 4 - 5 | 430 | 72 | |
| GenBank mRNA | BC109863.1 | - | 4 - 5 | 431 | 72 | |
More Information
Set Membership: |
|

APOD
Genomic Map