Assay Details
| Gene Symbol: FHL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076379.2 | NP_001069847.1 | 5 - 6 | 848 | 64 | |
| GenBank mRNA | BC110187.1 | - | 5 - 6 | 849 | 64 | |
More Information
Set Membership: |
|

FHL5
Genomic Map