Assay Details
| Gene Symbol: LYPD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076442.1 | NP_001069910.1 | 5 - 6 | 827 | 72 | |
| GenBank mRNA | BC109635.1 | - | 5 - 6 | 827 | 72 | |
More Information
Set Membership: |
|

LYPD4
Genomic Map