Assay Details
| Gene Symbol: TEPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302673.1 | NP_001289602.1 | 6 - 7 | 730 | 56 | |
| GenBank mRNA | BC110152.1 | - | 5 - 6 | 667 | 56 | |
More Information
Set Membership: |
|

TEPP
Genomic Map