Assay Details
Gene Symbol: KASH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076946.2 | NP_001070414.1 | 12 - 13 | 1220 | 85 | |
GenBank mRNA | BC111305.1 | - | 12 - 13 | 1221 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
