Assay Details
Gene Symbol: KASH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076946.2 | NP_001070414.1 | 2 - 3 | 273 | 94 | |
GenBank mRNA | BC111305.1 | - | 2 - 3 | 274 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC111305.1, NM_001076946.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
