Assay Details
| Gene Symbol: TUBB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077127.1 | NP_001070595.1 | 2 - 3 | 231 | 59 | |
| GenBank mRNA | BC111295.1 | - | 2 - 3 | 231 | 59 | |
More Information
Set Membership: |
|

TUBB3
Genomic Map