Assay Details
| Gene Symbol: COX6A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077831.2 | NP_001071299.1 | 1 - 2 | 120 | 77 | |
| GenBank mRNA | BC114182.1 | - | 1 - 2 | 121 | 77 | |
More Information
Set Membership: |
|

COX6A1
Genomic Map