Assay Details
| Gene Symbol: DMRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078060.1 | NP_001071528.1 | 4 - 5 | 1056 | 102 | |
| GenBank mRNA | BC109527.1 | - | 4 - 5 | 1056 | 102 | |
| FJ790227.1 | - | 4 - 5 | 919 | 102 | ||
More Information
Set Membership: |
|

DMRT1
Genomic Map