Assay Details
| Gene Symbol: TMIGD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079645.3 | NP_001073113.2 | 3 - 4 | 311 | 76 | |
| NM_001302767.1 | NP_001289696.1 | 2 - 3 | 723 | 76 | ||
| GenBank mRNA | BC109599.1 | - | 3 - 4 | 76 | ||
| DT839214.1 | - | 3 - 4 | 283 | 76 | ||
| DT846942.1 | - | 3 - 4 | 312 | 76 | ||
| DT901703.1 | - | 2 - 3 | 633 | 76 | ||
More Information
Set Membership: |
|

TMIGD3
Genomic Map