Assay Details
| Gene Symbol: NR1I3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079768.2 | NP_001073236.1 | 2 - 3 | 184 | 60 | |
| GenBank mRNA | AY789649.1 | - | 2 - 3 | 158 | 60 | |
| BC112658.1 | - | 2 - 3 | 185 | 60 | ||
More Information
Set Membership: |
|

NR1I3
Genomic Map