Assay Details
| Gene Symbol: PHYHIPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079798.1 | NP_001073266.1 | 1 - 2 | 241 | 90 | |
| GenBank mRNA | BC109692.1 | - | 1 - 2 | 241 | 90 | |
More Information
Set Membership: |
|

PHYHIPL
Genomic Map