Assay Details
Gene Symbol: LRRC46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079799.1 | NP_001073267.1 | 3 - 4 | 519 | 78 | |
GenBank mRNA | BC112534.1 | - | 3 - 4 | 519 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
