Assay Details
Gene Symbol: MED29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080316.2 | NP_001073785.1 | 2 - 3 | 287 | 79 | |
GenBank mRNA | BC126813.1 | - | 2 - 3 | 288 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
