Assay Details
| Gene Symbol: ANAPC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080357.2 | NP_001073826.1 | 5 - 6 | 798 | 91 | |
| GenBank mRNA | BC110224.1 | - | 5 - 6 | 799 | 91 | |
More Information
Set Membership: |
|

ANAPC10
Genomic Map