Assay Details
| Gene Symbol: H2BC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081742.1 | NP_001075211.1 | 1 - 2 | 497 | 78 | |
| GenBank mRNA | BC109769.1 | - | 1 - 2 | 497 | 78 | |
More Information
Set Membership: |
|

H2BC15
Genomic Map