Assay Details
Gene Symbol: CCKBR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174262.3 | NP_776687.2 | 1 - 2 | 256 | 116 | |
GenBank mRNA | BC112800.1 | - | 1 - 2 | 256 | 116 | |
S83090.1 | - | 1 - 2 | 178 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
