Assay Details
| Gene Symbol: FUNDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174338.4 | NP_776763.1 | 4 - 5 | 518 | 88 | |
| GenBank mRNA | AF375478.2 | - | 4 - 5 | 500 | 88 | |
| BC109698.1 | - | 4 - 5 | 519 | 88 | ||
More Information
Set Membership: |
|

FUNDC2
Genomic Map