Assay Details
Gene Symbol: TIMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174473.4 | NP_776898.2 | 1 - 2 | 402 | 60 | |
GenBank mRNA | AF144765.1 | - | 1 - 2 | 93 | 60 | |
BC112663.1 | - | 1 - 2 | 403 | 60 | ||
BC142426.1 | - | 1 - 2 | 361 | 60 | ||
U77588.1 | - | 1 - 2 | 452 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
