Assay Details
| Gene Symbol: STAT5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174617.4 | NP_777042.2 | 16 - 17 | 2162 | 78 | |
| GenBank mRNA | AJ237933.1 | - | 15 - 16 | 2079 | 78 | |
| BC112568.1 | - | 16 - 17 | 2163 | 78 | ||
More Information
Set Membership: |
|

STAT5B
Genomic Map