Assay Details
| Gene Symbol: RPLP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174788.4 | NP_777213.1 | 241 | 71 | ||
| GenBank mRNA | BC110143.1 | - | 242 | 71 | ||
| DQ347616.1 | - | 3 - 4 | 238 | 71 | ||
| U17836.1 | - | 3 - 4 | 212 | 71 | ||
More Information
Set Membership: |
|

RPLP2
Genomic Map