Assay Details
| Gene Symbol: NDUFS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302669.1 | NP_001289598.1 | 5 - 6 | 444 | 62 | |
| GenBank mRNA | BC109906.1 | - | 5 - 6 | 62 | ||
| DT719975.1 | - | 5 - 6 | 418 | 62 | ||
| DV906691.1 | - | 5 - 6 | 445 | 62 | ||
| M58717.1 | - | 5 - 6 | 422 | 62 | ||
More Information
Set Membership: |
|

NDUFS8
Genomic Map