Assay Details
Gene Symbol: ATP5IF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175816.3 | NP_787010.1 | 1 - 2 | 90 | 54 | |
GenBank mRNA | BC111645.1 | - | 1 - 2 | 93 | 54 | |
M22559.1 | - | 1 - 2 | 94 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
