Assay Details
| Gene Symbol: FABP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175817.3 | NP_787011.1 | 371 | 71 | ||
| GenBank mRNA | BC111622.1 | - | 372 | 71 | ||
| X86904.1 | - | 336 | 71 | |||
More Information
Set Membership: |
|

FABP1
Genomic Map