Assay Details
| Gene Symbol: USF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001161.2 | NP_001001161.1 | 10 - 11 | 960 | 67 | |
| GenBank mRNA | AY241931.1 | - | 8 - 9 | 845 | 67 | |
| BC114684.1 | - | 10 - 11 | 960 | 67 | ||
More Information
Set Membership: |
|

USF1
Genomic Map