Assay Details
| Gene Symbol: GFOD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014863.2 | NP_001014863.1 | 3 - 4 | 302 | 113 | |
| GenBank mRNA | BT025419.1 | - | 3 - 4 | 302 | 113 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001014863.2 transcripts.
Set Membership: |
|

GFOD2
Genomic Map