Assay Details
| Gene Symbol: GFOD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014863.2 | NP_001014863.1 | 4 - 5 | 646 | 66 | |
| GenBank mRNA | BC123696.1 | - | 2 - 3 | 458 | 66 | |
| BT021179.1 | - | 2 - 3 | 448 | 66 | ||
| BT025419.1 | - | 4 - 5 | 646 | 66 | ||
More Information
Set Membership: |
|

GFOD2
Genomic Map