Assay Details
| Gene Symbol: PIPOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014878.2 | NP_001014878.2 | 2 - 3 | 343 | 68 | |
| GenBank mRNA | BC114006.1 | - | 2 - 3 | 343 | 68 | |
| BT020944.1 | - | 2 - 3 | 338 | 68 | ||
More Information
Set Membership: |
|

PIPOX
Genomic Map