Assay Details
| Gene Symbol: DGUOK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014888.3 | NP_001014888.2 | 4 - 5 | 618 | 65 | |
| GenBank mRNA | BC112576.1 | - | 4 - 5 | 619 | 65 | |
| BT021046.1 | - | 4 - 5 | 599 | 65 | ||
More Information
Set Membership: |
|

DGUOK
Genomic Map