Assay Details
| Gene Symbol: CD53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034232.2 | NP_001029404.1 | 5 - 6 | 525 | 59 | |
| GenBank mRNA | BC113322.1 | - | 5 - 6 | 525 | 59 | |
| BT021574.1 | - | 5 - 6 | 493 | 59 | ||
More Information
Set Membership: |
|

CD53
Genomic Map