Assay Details
| Gene Symbol: SPEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034422.2 | NP_001029594.1 | 2 - 3 | 386 | 79 | |
| GenBank mRNA | BC118201.1 | - | 2 - 3 | 386 | 79 | |
| BT021696.1 | - | 2 - 3 | 367 | 79 | ||
More Information
Set Membership: |
|

SPEF1
Genomic Map