Assay Details
Gene Symbol: PBLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045886.2 | NP_001039351.1 | 2 - 3 | 221 | 79 | |
GenBank mRNA | BC105465.1 | - | 2 - 3 | 222 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
