Assay Details
Gene Symbol: PBLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045886.2 | NP_001039351.1 | 4 - 5 | 424 | 65 | |
GenBank mRNA | BC105465.1 | - | 4 - 5 | 425 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
