Assay Details
Gene Symbol: PLPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045890.1 | NP_001039355.1 | 1 - 2 | 116 | 89 | |
GenBank mRNA | BC113292.1 | - | 1 - 2 | 116 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
